Methyl-2,5-difluoro-4-nitrobenzoate 95%
Supplier: Thermo Scientific Chemicals
H63472-03
H63472-06
AAH63472-03EA
221.2
CAD
AAH63472-03
AAH63472-06
AAH63472-MD
Methyl-2,5-difluoro-4-nitrobenzoate 95%
Methyl 2,5-difluoro-4-nitrobenzoate
Methyl 2,5-difluoro-4-nitrobenzoate, 95%
Purity: 95%
CAS No.: 924868-81-5
Molecular Formula: C8H5F2NO4
Formula Weight: 217.13
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD08689670
Formula:
C₈H₅F₂NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD08689670 CAS Number: 924868-81-5 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...