3-Hydroxyisonicotinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63465-03
H63465-06
AAH63465-03EA
49.14
CAD
AAH63465-03
AAH63465-06
3-Hydroxyisonicotinic acid 98%
3-Hydroxyisonicotinic acid
3-Hydroxypyridine-4-carboxylic acid, 98%
Purity: 98%
CAS No.: 10128-71-9
Molecular Formula: C6H5NO3
Formula Weight: 139.11
Physical Form: Solid
Beilstein Reference: 383677
MDL No.: MFCD00234165
Formula:
C₆H₅NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00234165 CAS Number: 10128-71-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...