H-D-Glu(OtBu)-OH 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
D-Glutamic acid 5-tert-butyl ester
H63463-03
H63463-06
H63463-14
AAH63463-03EA
54.98
CAD
AAH63463-03
AAH63463-06
AAH63463-14
H-D-Glu(OtBu)-OH 95%
H-D-Glu(OtBu)-OH
D-Glutamic acid 5-tert-butyl ester, 95%
Purity: 95%
CAS No.: 45125-00-6
Molecular Formula: C9H17NO4
Formula Weight: 203.24
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00153437
Formula:
C₉H₁₇NO₄ MW: 203.238 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00153437 CAS Number: 45125-00-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...