N-[(9H-Fluoren-9-ylmethoxy)carbonyl]anthranilic acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-2-Abz-OH, 2-(Fmoc-amino)benzoic Acid, 2-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]benzoic Acid, N-Fmoc-anthranilic Acid
H63459-06
H63459-14
AAH63459-06EA
95.27
CAD
AAH63459-06
AAH63459-14
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]anthranilic acid 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]anthranilic acid
2-(Fmoc-amino)benzoic acid, 98%
Purity: 98%
CAS No.: 150256-42-1
Molecular Formula: C22H17NO4
Formula Weight: 359.37
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 216-218° dec
Beilstein Reference: 7659726
MDL No.: MFCD00235883
Formula:
C₂₂H₁₇NO₄ Melting Pt: 216…218 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00235883 CAS Number: 150256-42-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...