7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

H63456-03 H63456-06 H63456-MD
AAH63456-03EA 268.6 CAD
AAH63456-03 AAH63456-06 AAH63456-MD
7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%
7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride

7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%


Purity: 98% CAS No.: 73075-45-3 Molecular Formula: C9H10ClN.HCl Formula Weight: 204.10 Physical Form: Solid MDL No.: MFCD08461076

Formula: C₉H₁₁Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD08461076
CAS Number: 73075-45-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR