Fmoc-D-allo-isoleucine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-D-allo-Ile-OH, N-Fmoc-D-allo-isoleucine

H63455-03 H63455-06
AAH63455-03EA 297.04 CAD
AAH63455-03 AAH63455-06 AAH63455-MD
Fmoc-D-allo-isoleucine 98%
Fmoc-D-allo-isoleucine

N-Fmoc-D-allo-isoleucine, 98%


Purity: 98% CAS No.: 118904-37-3 Molecular Formula: C21H23NO4 Formula Weight: 353.41 Storage Temperature: 2°C to 8°C Physical Form: Solid MDL No.: MFCD00077062

Formula: C₂₁H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00077062
CAS Number: 118904-37-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR