N-Acetyl-D-alanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Ac-D-Ala-OH, Ac-D-Ala-OH
H63450-03
H63450-06
AAH63450-03EA
59.25
CAD
AAH63450-03
AAH63450-06
N-Acetyl-D-alanine 98%
N-Acetyl-D-alanine
N-Acetyl-D-alanine, 98%
Purity: 98%
CAS No.: 19436-52-3
Molecular Formula: C5H9NO3
Formula Weight: 131.13
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00063133
Formula:
C₅H₉NO₃ MW: 131.131 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00063133 CAS Number: 19436-52-3 EINECS: 243-066-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...