Fmoc-4-chloro-L-phenylalanine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-4-chloro-Phe-OH, N-Fmoc-4-chloro-L-phenylalanine, Fmoc-Phe(4-Cl)-OH

H63448-03 H63448-06
AAH63448-03EA 149.47 CAD
AAH63448-03 AAH63448-06 AAH63448-MD
Fmoc-4-chloro-L-phenylalanine 98%
Fmoc-4-chloro-L-phenylalanine

N-Fmoc-4-chloro-L-phenylalanine, 98%


Purity: 98% CAS No.: 175453-08-4 Molecular Formula: C24H20ClNO4 Formula Weight: 421.87 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 9437838 MDL No.: MFCD00080281

Formula: C₂₄H₂₀ClNO₄
MW: 421.877 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00080281
CAS Number: 175453-08-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR