Ethyl-4-nitro-1H-indole-2-carboxylate 97%
Supplier: Thermo Scientific Chemicals
H63440-03
H63440-MD
AAH63440-03EA
1045.96
CAD
AAH63440-03
AAH63440-MD
Ethyl-4-nitro-1H-indole-2-carboxylate 97%
Ethyl-4-nitro-1H-indole-2-carboxylate
Ethyl 4-nitroindole-2-carboxylate, 97%
Purity: 97%
CAS No.: 4993-93-5
Molecular Formula: C11H10N2O4
Formula Weight: 234.21
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Yellow
MDL No.: MFCD06795512
Formula:
C₁₁H₁₀N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD06795512 CAS Number: 4993-93-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...