4-(Trifluoromethylthio)phenylacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-[(Trifluoromethyl)sulfanyl]phenylacetic acid, 4-(Carboxymethyl)phenyl trifluoromethyl sulfide, {4-[(Trifluoromethyl)sulfanyl]phenyl}acetic acid, 2-(4-((Trifluoromethyl)thio)phenyl)acetic acid, 4-(Carboxymethyl)phenyl trifluoromethyl sulphide, {4-[(Trifluoromethyl)sulphanyl]phenyl}acetic acid, 4-[(Trifluoromethyl)sulphanyl]phenylacetic acid

H63432-03 H63432-06
AAH63432-03EA 109.18 CAD
AAH63432-03 AAH63432-06 AAH63432-MD
4-(Trifluoromethylthio)phenylacetic acid 98%
4-(Trifluoromethylthio)phenylacetic acid

4-(Trifluoromethylthio)phenylacetic acid, 98%


Purity: 98% CAS No.: 243977-23-3 Molecular Formula: C9H7F3O2S Formula Weight: 236.21 Physical Form: Solid MDL No.: MFCD00236351

Formula: C₉H₇F₃O₂S
MW: 236.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236351
CAS Number: 243977-23-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR