Boc-O-benzyl-D-tyrosine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Boc-O-benzyl-D-tyrosine, Boc-D-Tyr(Bzl)-OH
H63427-06
H63427-14
AAH63427-03EA
45.95
CAD
AAH63427-03
AAH63427-06
AAH63427-14
Boc-O-benzyl-D-tyrosine 95%
Boc-O-benzyl-D-tyrosine
N-Boc-O-benzyl-D-tyrosine, 95%
Purity: 95%
CAS No.: 63769-58-4
Molecular Formula: C21H25NO5
Formula Weight: 371.43
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00038249
Formula:
C₂₁H₂₅NO₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD00038249 CAS Number: 63769-58-4 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...