2-chloro-5-iodopyrimidine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63423-03
H63423-06
AAH63423-03EA
111.86
CAD
AAH63423-03
AAH63423-06
AAH63423-MD
2-chloro-5-iodopyrimidine 95%
2-chloro-5-iodopyrimidine
2-Chloro-5-iodopyrimidine, 95%
Purity: 95%
CAS No.: 32779-38-7
Molecular Formula: C4H2ClIN2
Formula Weight: 240.43
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Off-white
MDL No.: MFCD08062390
Formula:
C₄H₂ClIN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD08062390 CAS Number: 32779-38-7 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...