D-Methionine methyl ester hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-D-Met-OMe·HCl, H-D-Met-OMe.HCl
H63422-06
H63422-14
H63422-22
AAH63422-06EA
52.14
CAD
AAH63422-06
AAH63422-14
AAH63422-22
D-Methionine methyl ester hydrochloride 98%
D-Methionine methyl ester hydrochloride
D-Methionine methyl ester hydrochloride, 98%
Purity: 98%
CAS No.: 69630-60-0
Molecular Formula: C6H13NO2S.HCl
Formula Weight: 199.7
Physical Form: Solid
MDL No.: MFCD00070385
Formula:
C₆H₁₃NO₂S·HCl MW: 199.7 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070385 CAS Number: 69630-60-0 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...