L-Alanine-4-nitroanilide hydrochloride 98%
Supplier: Thermo Scientific Chemicals
H63393-03
H63393-06
H63393-MD
AAH63393-03EA
164.32
CAD
AAH63393-03
AAH63393-06
AAH63393-MD
L-Alanine-4-nitroanilide hydrochloride 98%
L-Alanine-4-nitroanilide hydrochloride
L-Alanine 4-nitroanilide hydrochloride, 98%
Purity: 98%
CAS No.: 31796-55-1
Molecular Formula: C9H11N3O3.HCl
Formula Weight: 245.66
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Pale yellow
Beilstein Reference: 6303731
MDL No.: MFCD00039088
Formula:
C₉H₁₂ClN₃O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00039088 CAS Number: 31796-55-1 EINECS: 250-811-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...