Indole-4-carbonitrile 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Cyano-1H-indole, 1H-Indole-4-carbonitrile, 4-Cyanoindole

H63391-03 H63391-06
AAH63391-03EA 46.29 CAD
AAH63391-03 AAH63391-06
Indole-4-carbonitrile 99%
Indole-4-carbonitrile

4-Cyanoindole, 99%


Purity: 99% CAS No.: 16136-52-0 Molecular Formula: C9H6N2 Formula Weight: 142.16 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 120-125° MDL No.: MFCD00152045

Formula: C₉H₆N₂
Melting Pt: 120…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00152045
CAS Number: 16136-52-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR