Methyl-2-methylpropionimidate hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl isobutyrimidate hydrochloride
H63386-03
H63386-06
H63386-MD
AAH63386-03EA
98.28
CAD
AAH63386-03
AAH63386-06
AAH63386-MD
Methyl-2-methylpropionimidate hydrochloride 97%
Methyl-2-methylpropionimidate hydrochloride
Methyl 2-methylpropionimidate hydrochloride, 97%
Purity: 97%
CAS No.: 39739-60-1
Molecular Formula: C5H11NO.HCl
Formula Weight: 137.61
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White
MDL No.: MFCD03844704
Formula:
C₅H₁₂ClNO MW: 137.607 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03844704 CAS Number: 39739-60-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...