D-2-Cyclohexylglycine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-alpha-Aminocyclohexylacetic Acid, H-D-Chg-OH

H63381-03 H63381-06
AAH63381-03EA 63.52 CAD
AAH63381-03 AAH63381-06 AAH63381-14
D-2-Cyclohexylglycine 95%
D-2-Cyclohexylglycine

D-(-)-2-Cyclohexylglycine, 95%


Purity: 95% CAS No.: 14328-52-0 Molecular Formula: C8H15NO2 Formula Weight: 157.21 Physical Form: Solid Beilstein Reference: 3196806 MDL No.: MFCD01311678

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01311678
CAS Number: 14328-52-0

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR