Fmoc-Glu(OMe)-OH 97%

Supplier: Thermo Scientific Chemicals

H63358-06 H63358-14
AAH63358-06EA 80.42 CAD
AAH63358-06 AAH63358-14 AAH63358-22
Fmoc-Glu(OMe)-OH 97%
Fmoc-Glu(OMe)-OH

N-Fmoc-L-glutamic acid 5-methyl ester, 97%


Purity: 97% CAS No.: 145038-50-2 Molecular Formula: C21H21NO6 Formula Weight: 383.39 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD01076132

Formula: C₂₁H₂₁NO₆
Storage Temperature: Ambient
MDL Number: MFCD01076132
CAS Number: 145038-50-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR