Fmoc-Orn(Alloc)-OH 95%
Supplier: Thermo Scientific Chemicals
H63331-06
H63331-14
AAH63331-03EA
53.53
CAD
AAH63331-03
AAH63331-06
AAH63331-14
Fmoc-Orn(Alloc)-OH 95%
Fmoc-Orn(Alloc)-OH
N^d-Allyloxycarbonyl-N^a-Fmoc-L-ornithine, 95%
Purity: 95%
CAS No.: 147290-11-7
Molecular Formula: C24H26N2O6
Formula Weight: 438.47
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00798637
Formula:
C₂₄H₂₆N₂O₆ Storage Temperature: Refrigerator |
MDL Number:
MFCD00798637 CAS Number: 147290-11-7 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...