(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride 98%
Supplier: Thermo Scientific Chemicals
H63325-03
H63325-06
AAH63325-03EA
83.11
CAD
AAH63325-03
AAH63325-06
AAH63325-MD
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride 98%
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride
D-Phenylalanine tert-butyl ester hydrochloride, 98%
Purity: 98%
CAS No.: 3403-25-6
Molecular Formula: C13H19NO2.HCl
Formula Weight: 257.76
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00153455
Formula:
C13H20ClNO2 Storage Temperature: Refrigerator |
MDL Number:
MFCD00153455 CAS Number: 3403-25-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...