H-d-glu-otbu 98%

Supplier: Thermo Scientific Chemicals

Synonyms: D-Glutamic acid 1-tert-butyl ester, H-D-Glu-OtBu

H63318-03 H63318-06
AAH63318-03EA 118.18 CAD
AAH63318-03 AAH63318-06 AAH63318-MD
H-d-glu-otbu 98%
H-d-glu-otbu

D-Glutamic acid 1-tert-butyl ester, 98%


Purity: 98% CAS No.: 25456-76-2 Molecular Formula: C9H17NO4 Formula Weight: 203.24 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD09842819

Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09842819
CAS Number: 25456-76-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR