3-(2-Thienyl)-L-alanine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63316-03
H63316-06
AAH63316-03EA
254.38
CAD
AAH63316-03
AAH63316-06
AAH63316-MD
3-(2-Thienyl)-L-alanine 95%
3-(2-Thienyl)-L-alanine
3-(2-Thienyl)-L-alanine, 95%
Purity: 95%
CAS No.: 22951-96-8
Molecular Formula: C7H9NO2S
Formula Weight: 171.21
Physical Form: Solid
Appearance: Off-white
Melting Point: 255-263° dec
Beilstein Reference: 82872
Optical Rotation: -31°(c=1 in water)
MDL No.: MFCD00064335
Formula:
C₇H₉NO₂S Melting Pt: 255…263 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064335 CAS Number: 22951-96-8 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...