2-(Trifluoromethyl)isonicotinic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Trifluoromethyl)pyridine-4-carboxylic acid

H63315-03 H63315-MD
AAH63315-03EA 360.24 CAD
AAH63315-03 AAH63315-MD
2-(Trifluoromethyl)isonicotinic acid 97%
2-(Trifluoromethyl)isonicotinic acid

2-(Trifluoromethyl)pyridine-4-carboxylic acid, 97%


Purity: 97% CAS No.: 131747-41-6 Molecular Formula: C7H4F3NO2 Formula Weight: 191.11 Physical Form: Solid Appearance: Off-white Melting Point: 217-223° MDL No.: MFCD07774132

Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Melting Pt: 217…223 °C
Storage Temperature: Ambient
MDL Number: MFCD07774132
CAS Number: 131747-41-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR