Boc-N-methyl-L-serine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-N-Me-Ser-OH, N-Boc-N-methyl-L-serine

H63314-03 H63314-06
AAH63314-03EA 176.39 CAD
AAH63314-03 AAH63314-06 AAH63314-MD
Boc-N-methyl-L-serine 98%
Boc-N-methyl-L-serine

N-Boc-N-methyl-L-serine, 98%


Purity: 98% CAS No.: 101772-29-6 Molecular Formula: C9H17NO5 Formula Weight: 219.23 Physical Form: Solid Melting Point: 83-87° Beilstein Reference: 8685334 Optical Rotation: -33°(c=1 in methanol) MDL No.: MFCD00037249

Formula: C₉H₁₇NO₅
Melting Pt: 83…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00037249
CAS Number: 101772-29-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR