1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: O-(4-Methoxybenzyl)hydroxylamine hydrochloride

H63312-03 H63312-06
AAH63312-03EA 106.49 CAD
AAH63312-03 AAH63312-06
1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride 98%
1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride

O-(4-Methoxybenzyl)hydroxylamine hydrochloride, 98%


Purity: 98% CAS No.: 21038-22-2 Molecular Formula: C8H11NO2 Formula Weight: 153.18 Physical Form: Solid Appearance: White MDL No.: MFCD01114582

Formula: C₈H₁₂ClNO₂
MW: 189.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01114582
CAS Number: 876-33-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR