(3,5-Dichlorophenyl)acetic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63293-03
H63293-06
AAH63293-03EA
104.12
CAD
AAH63293-03
AAH63293-06
AAH63293-MD
(3,5-Dichlorophenyl)acetic acid 95%
(3,5-Dichlorophenyl)acetic acid
3,5-Dichlorophenylacetic acid, 95%
Purity: 95%
CAS No.: 51719-65-4
Molecular Formula: C8H6Cl2O2
Formula Weight: 205.03
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD01764671
Formula:
C₈H₆Cl₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD01764671 CAS Number: 51719-65-4 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...