N-Fmoc-4-iodo-D-phenylalanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-D-Phe(4-I)-OH
H63292-03
H63292-06
AAH63292-03EA
94.64
CAD
AAH63292-03
AAH63292-06
AAH63292-MD
N-Fmoc-4-iodo-D-phenylalanine 98%
N-Fmoc-4-iodo-D-phenylalanine
N-Fmoc-4-iodo-D-phenylalanine, 98%
Purity: 98%
CAS No.: 205526-29-0
Molecular Formula: C24H20INO4
Formula Weight: 513.32
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Light Sensitive
Beilstein Reference: 9159551
MDL No.: MFCD00672557
Formula:
C₂₄H₂₀INO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00672557 CAS Number: 205526-29-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...