2-Bromobenzothiazole 99%

Supplier: Thermo Scientific Chemicals
Warning

H63286-03 H63286-06 H63286-14
AAH63286-03EA 78.05 CAD
AAH63286-03 AAH63286-06 AAH63286-14
2-Bromobenzothiazole 99%

2-Bromobenzothiazole


Purity: 99% CAS No.: 2516-40-7 Molecular Formula: C7H4BrNS Formula Weight: 214.08 Flash Point: 110°(230°F) Physical Form: Crystalline solid Appearance: Pale yellow Melting Point: 39-43° MDL No.: MFCD02681887

Formula: C₇H₄BrNS
Melting Pt: 39…43 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD02681887
CAS Number: 2516-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR