(S)-2-Amino-4-oxo-4-(tritylamino)butanoic acid hydrate 98%
Supplier: Thermo Scientific Chemicals
H63285-03
H63285-06
AAH63285-03EA
60.2
CAD
AAH63285-03
AAH63285-06
AAH63285-MD
(S)-2-Amino-4-oxo-4-(tritylamino)butanoic acid hydrate 98%
(S)-2-Amino-4-oxo-4-(tritylamino)butanoic acid hydrate
N^y-Trityl-L-asparagine hydrate, 98%
Purity: 98%
CAS No.: 132388-58-0
Molecular Formula: C23H22N2O3.xH2O
Formula Weight: 392.45(anhy)
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00153422
Formula:
C₂₃H₂₂N₂O₃·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00153422 CAS Number: 210824-10-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...