h-d-Tyr-otbu 98%
Supplier: Thermo Scientific Chemicals
H63274-03
H63274-06
AAH63274-03EA
55.3
CAD
AAH63274-03
AAH63274-06
h-d-Tyr-otbu 98%
h-d-Tyr-otbu
D-Tyrosine tert-butyl ester, 98%
Purity: 98%
CAS No.: 87553-74-0
Molecular Formula: C13H19NO3
Formula Weight: 237.14
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00191035
Formula:
C₁₃H₁₉NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00191035 CAS Number: 87553-74-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...