Fmoc-Met(O)-OH 98%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Fmoc-L-methionine sulphoxide, N-Fmoc-L-methionine sulfoxide

H63271-03 H63271-06
AAH63271-03EA 57.83 CAD
AAH63271-03 AAH63271-06
Fmoc-Met(O)-OH 98%
Fmoc-Met(O)-OH

N-Fmoc-L-methionine sulfoxide, 98%


Purity: 98% CAS No.: 76265-70-8 Molecular Formula: C20H21NO5S Formula Weight: 387.45 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD00077064

Formula: C₂₀H₂₁NO₅S
MW: 387.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077064
CAS Number: 76265-70-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR