Methyl-4-fluoro-2-methylbenzoate 98%

Supplier: Thermo Scientific Chemicals

H63268-06 H63268-14 H63268-22
AAH63268-06EA 68.73 CAD
AAH63268-06 AAH63268-14 AAH63268-22
Methyl-4-fluoro-2-methylbenzoate 98%
Methyl 4-fluoro-2-methylbenzoate

Methyl 4-fluoro-2-methylbenzoate, 98%


Purity: 98% CAS No.: 174403-69-1 Molecular Formula: C9H9FO2 Formula Weight: 168.16 Physical Form: Liquid Appearance: Colorless MDL No.: MFCD06203790

Formula: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD06203790
CAS Number: 174403-69-1

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR