H-Asp(OtBu)-OMe·HCl 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride, H-Asp(OtBu)-OMe.HCl
H63257-03
H63257-06
AAH63257-03EA
114.55
CAD
AAH63257-03
AAH63257-06
AAH63257-MD
H-Asp(OtBu)-OMe·HCl 98%
H-Asp(OtBu)-OMe·HCl
L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride, 98%
Purity: 98%
CAS No.: 2673-19-0
Molecular Formula: C9H17NO4.HCl
Formula Weight: 239.7
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 6005591
MDL No.: MFCD00153427
Formula:
C9H18ClNO4 MW: 239.7 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00153427 CAS Number: 2673-19-0 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...