(R)-Piperidine-2-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63255-06
H63255-14
H63255-22
AAH63255-06EA
60.67
CAD
AAH63255-06
AAH63255-14
AAH63255-22
(R)-Piperidine-2-carboxylic acid 97%
(R)-Piperidine-2-carboxylic acid
D-Pipecolinic acid, 97%
Purity: 97%
CAS No.: 1723-00-8
Molecular Formula: C6H11NO2
Formula Weight: 129.16
Physical Form: Solid
Melting Point: 277° dec
Beilstein Reference: 81094
Optical Rotation: +27°(c=1 in water)
MDL No.: MFCD00064346
Formula:
C₆H₁₁NO₂ Melting Pt: 277 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064346 CAS Number: 1723-00-8 EINECS: 217-024-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...