N-Carbobenzoxy-L-valine succinimidyl ester 98%
Supplier: Thermo Scientific Chemicals
H63253-06
H63253-14
AAH63253-06EA
61.15
CAD
AAH63253-06
AAH63253-14
N-Carbobenzoxy-L-valine succinimidyl ester 98%
N-Carbobenzoxy-L-valine succinimidyl ester
N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, 98%
Purity: 98%
CAS No.: 3496-11-5
Molecular Formula: C17H20N2O6
Formula Weight: 348.35
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 114-118°
Beilstein Reference: 1555287
Optical Rotation: -25°(c=1 in dioxane)
MDL No.: MFCD00053547
Formula:
C₁₇H₂₀N₂O₆ Melting Pt: 114…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053547 CAS Number: 3496-11-5 EINECS: 222-493-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...