2-Fluoro-3-nitrobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63235-03
H63235-06
AAH63235-03EA
59.72
CAD
AAH63235-03
AAH63235-06
2-Fluoro-3-nitrobenzoic acid 97%
2-Fluoro-3-nitrobenzoic acid
2-Fluoro-3-nitrobenzoic acid, 97%
Purity: 97%
CAS No.: 317-46-4
Molecular Formula: C7H4FNO4
Formula Weight: 185.11
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD06797825
Formula:
C₇H₄FNO₄ MW: 185.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06797825 CAS Number: 317-46-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...