(2R,2'R)-Dimethyl-3,3'-disulfanediylbis(2-aminopropanoate) dihydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(H-Cystine-OMe)₂·2HCl, L-Cystine dimethyl ester dihydrochloride, (H-Cys-OMe)₂·2 HCl, (H-Cys-OMe)2.2HCl
H63234-03
H63234-06
H63234-14
AAH63234-03EA
42.82
CAD
AAH63234-03
AAH63234-06
AAH63234-14
(2R,2'R)-Dimethyl-3,3'-disulfanediylbis(2-aminopropanoate) dihydrochloride 98%
(2R,2'R)-Dimethyl-3,3'-disulfanediylbis(2-aminopropanoate) dihydrochloride
L-Cystine dimethyl ester dihydrochloride, 98%
Purity: 98%
CAS No.: 32854-09-4
Molecular Formula: C8H16N2O4S2.2HCl
Formula Weight: 341.28
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 182-183° dec
Beilstein Reference: 4727524
Optical Rotation: -36°(c=4 in methanol)
MDL No.: MFCD00012490
Formula:
C₈H₁₈Cl₂N₂O₄S₂ MW: 341.28 g/mol Melting Pt: 182…183 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012490 CAS Number: 32854-09-4 EINECS: 251-261-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...