2-Amino-3-(1H-indol-3-yl)propanamide 98%

Supplier: Thermo Scientific Chemicals

Synonyms: DL-Tryptophanamide

H63227-03 H63227-MD
AAH63227-03EA 211.72 CAD
AAH63227-03 AAH63227-06 AAH63227-MD
2-Amino-3-(1H-indol-3-yl)propanamide 98%
2-Amino-3-(1H-indol-3-yl)propanamide

DL-Tryptophanamide, 98%


Purity: 98% CAS No.: 6720-02-1 Molecular Formula: C11H13N3O Formula Weight: 203.24 Physical Form: Crystalline powder Appearance: Off-white MDL No.: MFCD03826622

Formula: C₁₁H₁₃N₃O
MW: 203.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03826622
CAS Number: 6720-02-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR