1-Methyl-L-aspartate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Asp-OMe, (S)-3-Amino-4-methoxy-4-oxobutanoic acid, L-Aspartic acid 1-methyl ester

H63221-03 H63221-06
AAH63221-03EA 53.37 CAD
AAH63221-03 AAH63221-06 AAH63221-MD
1-Methyl-L-aspartate 98%
1-Methyl-L-aspartate

L-Aspartic acid 1-methyl ester, 98%


Purity: 98% CAS No.: 17812-32-7 Molecular Formula: C5H9NO4 Formula Weight: 147.13 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 186° dec Beilstein Reference: 2080763 Optical Rotation: +36°(c=0.5 in water) MDL No.: MFCD00080289

Formula: C₅H₉NO₄
Melting Pt: 186 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00080289
CAS Number: 17812-32-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR