Ethyl-2-methylquinoline-6-carboxylate 97%
Supplier: Thermo Scientific Chemicals
H63213-03
H63213-06
AAH63213-03EA
89.43
CAD
AAH63213-03
AAH63213-06
Ethyl-2-methylquinoline-6-carboxylate 97%
Ethyl-2-methylquinoline-6-carboxylate
Ethyl 2-methylquinoline-6-carboxylate, 97%
Purity: 97%
CAS No.: 855763-77-8
Molecular Formula: C13H13NO2
Formula Weight: 215.25
Physical Form: Solid
MDL No.: MFCD09879697
Formula:
C₁₃H₁₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09879697 CAS Number: 855763-77-8 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...