N-Boc-guanidine 95%

Supplier: Thermo Scientific Chemicals
Warning

H63206-06 H63206-14
AAH63206-03EA 36.28 CAD
AAH63206-03 AAH63206-06 AAH63206-14
N-Boc-guanidine 95%
N-Boc-guanidine

N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.


Purity: 95% CAS No.: 219511-71-4 Molecular Formula: C6H13N3O2 Formula Weight: 159.19 Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: White Melting Point: 170-175° MDL No.: MFCD20547465

Formula: C₆H₁₃N₃O₂
Melting Pt: 170…175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20547465
CAS Number: 219511-71-4

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR