N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine 97%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Boc-3-(2-naphthyl)-L-alanine, (S)-alpha-(Boc-amino)-2-naphthalenepropionic Acid, (S)-alpha-tert-Butoxycarbonylamino-2-naphthalenepropionic Acid, Boc-2-Nal-OH

H63205-03 H63205-06
AAH63205-03EA 100.17 CAD
AAH63205-03 AAH63205-06 AAH63205-14
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine 97%
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine

N-Boc-3-(2-naphthyl)-L-alanine, 97%


Purity: 97% CAS No.: 58438-04-3 Molecular Formula: C18H21NO4 Formula Weight: 315.37 Physical Form: Solid Appearance: Off-white Melting Point: ca 95° Beilstein Reference: 4326467 Optical Rotation: +45°(c=1 in ethanol) MDL No.: MFCD00079671

Formula: C₁₈H₂₁NO₄
Melting Pt: ∼ 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00079671
CAS Number: 58438-04-3

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR