4-Ethyl-1H-pyrazol-3-amine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-4-ethyl-1H-pyrazole

H63199-03 H63199-06
AAH63199-03EA 306.52 CAD
AAH63199-03 AAH63199-06 AAH63199-MD
4-Ethyl-1H-pyrazol-3-amine 98%
4-Ethyl-1H-pyrazol-3-amine

3-Amino-4-ethyl-1H-pyrazole, 98%


Purity: 98% CAS No.: 43024-15-3 Molecular Formula: C5H9N3 Formula Weight: 111.15 Physical Form: Solid MDL No.: MFCD06797570

Formula: C₅H₉N₃
MW: 111.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797570
CAS Number: 43024-15-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR