(R)-Dimethyl-2-aminosuccinate hydrochloride 98%

Supplier: Thermo Scientific Chemicals

H63197-03 H63197-06
AAH63197-03EA 62.25 CAD
AAH63197-03 AAH63197-06
(R)-Dimethyl-2-aminosuccinate hydrochloride 98%
(R)-Dimethyl-2-aminosuccinate hydrochloride

D-Aspartic acid dimethyl ester hydrochloride, 98%


Purity: 98% CAS No.: 69630-50-8 Molecular Formula: C6H11NO4.HCl Formula Weight: 197.62 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 114-119° Optical Rotation: -14°(c=1 in methanol) MDL No.: MFCD00070384

Formula: C₆H₁₂ClNO₄
Melting Pt: 114…119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00070384
CAS Number: 69630-50-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR