Methyl-6-bromo-1H-indole-2-carboxylate 97%
Supplier: Thermo Scientific Chemicals
H63187-03
H63187-06
AAH63187-03EA
124.19
CAD
AAH63187-03
AAH63187-06
AAH63187-MD
Methyl-6-bromo-1H-indole-2-carboxylate 97%
Methyl-6-bromo-1H-indole-2-carboxylate
Methyl 6-bromoindole-2-carboxylate, 97%
Purity: 97%
CAS No.: 103858-53-3
Molecular Formula: C10H8BrNO2
Formula Weight: 254.08
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White
MDL No.: MFCD03094942
Formula:
C11H10BrNO2 Storage Temperature: Ambient |
MDL Number:
MFCD03094942 CAS Number: 372089-59-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...