3,4-Xylylacetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,4-Dimethylphenylacetic acid
H63178-03
H63178-06
AAH63178-03EA
74.58
CAD
AAH63178-03
AAH63178-06
3,4-Xylylacetic acid 98%
3,4-Xylylacetic acid
3,4-Dimethylphenylacetic acid, 98%
Purity: 98%
CAS No.: 17283-16-8
Molecular Formula: C10H12O2
Formula Weight: 164.2
Physical Form: Solid
MDL No.: MFCD02664684
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02664684 CAS Number: 17283-16-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...