tert-Butyl-2-(methylamino)acetate hydrochloride 97%
Supplier: Thermo Scientific Chemicals
H63176-06
H63176-14
AAH63176-06EA
124.19
CAD
AAH63176-06
AAH63176-14
tert-Butyl-2-(methylamino)acetate hydrochloride 97%
tert-Butyl-2-(methylamino)acetate hydrochloride
Sarcosine tert-butyl ester hydrochloride, 97%
Purity: 97%
CAS No.: 5616-81-9
Molecular Formula: C7H15NO2.HCl
Formula Weight: 181.66
Physical Form: Powder
Appearance: White
Melting Point: 137-141°
MDL No.: MFCD00080936
Formula:
C₇H₁₆ClNO₂ Melting Pt: 137…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00080936 CAS Number: 136088-69-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...