3-(Trifluoromethyl)isonicotinic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(Trifluoromethyl)pyridine-4-carboxylic acid

H63174-03 H63174-06
AAH63174-03EA 169.06 CAD
AAH63174-03 AAH63174-06 AAH63174-MD
3-(Trifluoromethyl)isonicotinic acid 95%
3-(Trifluoromethyl)isonicotinic acid

3-(Trifluoromethyl)pyridine-4-carboxylic acid, 95%


Purity: 95% CAS No.: 590371-38-3 Molecular Formula: C7H4F3NO2 Formula Weight: 191.11 Physical Form: Solid Appearance: Off-white MDL No.: MFCD08234940

Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08234940
CAS Number: 590371-38-3

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR