Boc-Phe-OH 95%
Supplier: Thermo Scientific Chemicals
H63164-06
H63164-14
AAH63164-03EA
36.72
CAD
AAH63164-03
AAH63164-06
AAH63164-14
Boc-Phe-OH 95%
Boc-Phe-OH
N-Boc-DL-phenylalanine, 95%
Purity: 95%
CAS No.: 4530-18-1
Molecular Formula: C14H19NO4
Formula Weight: 265.3
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00558969
Formula:
C₁₄H₁₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00558969 CAS Number: 4530-18-1 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...