N-Carbobenzoxy-D-methionine 98%

Supplier: Thermo Scientific Chemicals

H63152-06 H63152-14
AAH63152-03EA 38.66 CAD
AAH63152-03 AAH63152-06 AAH63152-14
N-Carbobenzoxy-D-methionine 98%
N-Carbobenzoxy-D-methionine

N-Benzyloxycarbonyl-D-methionine, 98%


Purity: 98% CAS No.: 28862-80-8 Molecular Formula: C13H17NO4S Formula Weight: 283.24 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 68-72° dec Beilstein Reference: 2058695 Optical Rotation: +26°(c=1 in methanol) MDL No.: MFCD00026043

Formula: C₁₃H₁₇NO₄S
Melting Pt: 68…72 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00026043
CAS Number: 28862-80-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR